missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012961
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012961 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012961 Supplier Merck Emd Millipore Supplier No. PHR2011100MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 133481-09-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H13 NO2
MDL Number MFCD04117948
Synonym (1-Cyanocyclohexyl)acetic acid; (1-CYANOCYCLOHEXYL)ACETIC ACID; 1-Cyanocyclohexaneacetic acid
Type Certified Reference Material
Quantity 100 mg
Formula Weight 167.21
Grade Certified Reference Material