missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 64744-50-9 |
| Physical Form | Neat |
| Linear Formula | C9H15 NO |
| MDL Number | MFCD00177938 |
| Melting Point | 84°C to 89°C |
| Synonym | 4,4-Pentamethylene-2-pyrrolidinone; 4,4-PENTAMETHYLENE-2-PYRROLIDONE; Gabapentin Impurity A; 3-Azaspiro[4.5 ]decan-2-one |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 153.22 |
| Grade | Certified Reference Material |