missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014829
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014829 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014829 Supplier Merck Emd Millipore Supplier No. PHR10491G

Gabapentin is an antiepileptic drug, which is a structural analog of γ-aminobu-tyric acid (GABA). It is used in the treatment of partial seizures in adults and children.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 60142-96-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C9H17 NO2
MDL Number MFCD00865286
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid; Neurontin
Type Certified Reference Material
Quantity 1 g
Formula Weight 171.24
Grade Certified Reference Material