Learn More
- Home
- Products
- Industrial Hygiene and Environmental Monitoring
- Gas and Vapor Detection
- Multigas Portable Detectors
- Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Catalog No. 170000077
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
Carbon Dioxide, LEL-I, Oxygen
Carbon Dioxide, Oxygen
Carbon Monoxide, Oxygen
Hydrogen Sulfide, Carbon Monoxide
Hydrogen Sulfide, Carbon Monoxide, Oxygen
Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, LEL-I, Oxygen, PID
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
LEL-I, Carbon Monoxide, Oxygen
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
LEL-I, Chlorine, Carbon Monoxide, Oxygen
LEL-I, Chlorine, Oxygen
LEL-I, Chlorine, Oxygen, Sulfur Dioxide
LEL-I, Oxygen
LEL-I, Oxygen, PID
LEL-I, PID
Methane, Carbon Dioxide
Methane, Carbon Dioxide, LEL-I, Oxygen
Methane, Carbon Dioxide, Oxygen
Methane, Carbon Monoxide
Methane, Hydrogen Sulfide, LEL-I, Oxygen
Methane, Hydrogen Sulfide, Oxygen
Methane, LEL-I, Carbon Monoxide, Oxygen
Methane, LEL-I, Chlorine, Oxygen
Methane, LEL-I, Oxygen
Methane, LEL-I, Oxygen, Carbon Monoxide - H
Methane, LEL-I, Oxygen, Sulfur Dioxide
Methane, Oxygen
Oxygen, PID
36 product options available for selection
| Catalog No. | Components Measured |
|---|---|
| 170000077 | Methane, Carbon Dioxide, Oxygen |
| 170000075 | Methane, Carbon Dioxide, LEL-I, Oxygen |
| 170000078 | Methane, Carbon Dioxide |
| 170000081 | Methane, Hydrogen Sulfide, LEL-I, Oxygen |
| 170000082 | Methane, Hydrogen Sulfide, Oxygen |
| 170000083 | Methane, LEL-I, Chlorine, Oxygen |
| 170000084 | Methane, LEL-I, Carbon Monoxide, Oxygen |
| 170000085 | Methane, LEL-I, Oxygen, Carbon Monoxide - H |
| 170000086 | Methane, LEL-I, Oxygen, Sulfur Dioxide |
| 170000087 | Methane, LEL-I, Oxygen |
| 170000089 | Methane, Carbon Monoxide |
| 170000090 | Methane, Oxygen |
| 170000093 | Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen |
| 170000094 | Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen |
| 170000095 | Carbon Dioxide, LEL-I, Oxygen |
| 170000096 | Carbon Dioxide, Oxygen |
| 170000098 | Carbon Dioxide, Oxygen |
| 170000104 | Hydrogen Sulfide, LEL-I, Chlorine, Oxygen |
| 170000105 | Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen |
| 170000106 | Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H |
| 170000107 | Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide |
| 170000108 | Hydrogen Sulfide, LEL-I, Oxygen, PID |
| 170000109 | Hydrogen Sulfide, LEL-I, Oxygen |
| 170000111 | Hydrogen Sulfide, Carbon Monoxide, Oxygen |
| 170000112 | Hydrogen Sulfide, Carbon Monoxide |
| 170000115 | Hydrogen Sulfide, Oxygen, Carbon Monoxide - H |
| 170000116 | LEL-I, Chlorine, Carbon Monoxide, Oxygen |
| 170000117 | LEL-I, Chlorine, Oxygen, Sulfur Dioxide |
| 170000118 | LEL-I, Chlorine, Oxygen |
| 170000119 | LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide |
| 170000120 | LEL-I, Carbon Monoxide, Oxygen |
| 170000121 | LEL-I, Oxygen, PID |
| 170000122 | LEL-I, Oxygen |
| 170000123 | LEL-I, PID |
| 170000128 | Carbon Monoxide, Oxygen |
| 170000132 | Oxygen, PID |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Specifications
| Components Measured | Methane, Carbon Dioxide, Oxygen |
| Type | Multigas Cartridge |
| Frequency | 900 MHz |