missing translation for 'onlineSavingsMsg'
Learn More

Antibiotic G-418 Sulfate Solution, Promega

Catalog No. PRV8091
Encompass
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8091 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV8091 Supplier Promega Corporation Supplier No. V8091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An aminoglycosidic antibiotic toxic to both prokaryotic and eukaryotic cells.

  • Available as a powder (5g) or as a liquid (20ml)
  • Liquid formulation requires no sterilization

Chemical Identifiers

CAS 108321-42-2 , 7732-18-5

Specifications

Physical Form Liquid
Color White
Quantity 20 mL
Grade Not Available
Recommended Storage −30 to +10°C