missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FZD9 Recombinant Rabbit Monoclonal Antibody (23GB5475)
GREENER_CHOICE

Catalog No. MA552862
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552862 100 μL
MA552863 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552862 Supplier Invitrogen™ Supplier No. MA552862
Only null left

Rabbit Recombinant Monoclonal Antibody

FZD9 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.

Specifications

Antigen FZD9
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5475
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FZD9
Gene Accession No. O00144
Gene Alias CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9
Gene Symbols FZD9
Host Species Rabbit
Immunogen A synthesized peptide derived from human Frizzled 9/CD349 (550-591AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less