missing translation for 'onlineSavingsMsg'
Learn More

FXN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102473 Supplier Thermo Fisher Scientific Supplier No. PA5102473
Only null left

FXN Polyclonal Antibody

FXN Polyclonal Antibody for Western Blot, IHC (P)

Specifications

Antigen FXN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FXN
Gene Accession No. D3ZYW7, O35943, Q16595
Gene Alias CyaY; d-FXN; DKEYP-118A9.1; FA; FARR; frataxin; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; FRDA; Friedreich ataxia; Friedreich ataxia protein; FXN; i-FXN; m56-FXN; m78-FXN; m81-FXN; RGD1565754; X25; zgc:158218
Gene Symbols FXN
Host Species Rabbit
Immunogen A synthesized peptide derived from human FXN(Accession Q16595), corresponding to amino acid residues L140-L190.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14297, 2395, 499335
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less