missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FxCycle™ Violet Stain

Catalog No. F10347
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10347 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10347 Supplier Invitrogen™ Supplier No. F10347

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FxCycle™ Violet Stain

FxCycle™ Violet stain (F10347) is used for flow cytometric analysis of DNA content in fixed cells. This dye is excited with a 405/407 nm laser with emission collected in the 450/50 BP filter or equilavent. This dye is DNA selective so RNAse is not required. With multicolor cell cycle studies possible using flow cytometry, it is often necessary to analyze DNA content on alternative lasers to preserve the common 488 nm laser and detection channels for other markers. Well suited for the popular violet laser line, FxCycle™ Violet stain is a good choice for DNA content analysis in multicolor cell cycle studies.

Related Links:

Specifications

Content And Storage Contains 5 vials of FxCycle™ violet stain (100 μg per vial). Store at 2-8°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type FxCycle™ Violet Stain
Form Liquid
Format Tube(s)
Product Line FxCycle, Molecular Probes
Quantity 1 kit
Shipping Condition Room Temperature
Sub Cellular Localization Nucleic Acids
Emission Wavelength 358/461
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.