missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FUT3 Monoclonal Antibody (1F8G5C7)
GREENER_CHOICE

Catalog No. FERMA548463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548463 Supplier Invitrogen™ Supplier No. MA548463

Mouse Monoclonal Antibody

FUT3 Monoclonal Antibody for Western Blot, Flow, ELISA

FUT3 comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This protein is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities.

Specifications

Antigen FUT3
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1F8G5C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene FUT3
Gene Accession No. P21217
Gene Alias 4-fucosyltransferase; alpha-(1,3/1,4)-fucosyltransferase; blood group Lewis alpha-4-fucosyltransferase; CD174; FT3B; Fucosyltransferase 3; fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group); fucosyltransferase 3 (Lewis blood group); fucosyltransferase III; FucT-III; Fuc-TV; FucT-V; FUT3; Galactoside 3(4)-L-fucosyltransferase; LE; Les; Lewis FT; Lewis Y
Gene Symbols FUT3
Host Species Mouse
Immunogen Purified recombinant fragment of human CD174 (AA: 199-361) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less