missing translation for 'onlineSavingsMsg'
Learn More

Fusidic acid, 98%, Thermo Scientific Chemicals

$592.73 - $592.73

Chemical Identifiers

CAS 6990-06-3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC458460050
View Documents
Thermo Scientific Chemicals
458460050
5 g
Each for $592.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6990-06-3

Specifications

6990-06-3
97.5% min. (HPLC)
5 g
Solubility in water: .. Other solubilities: soluble in methanol,chloroform
COC(C1=CC=CC=C1)C(=O)O
516.71
516.71
Fusidic acid
Authentic
C31H48O6
methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
DIWVBIXQCNRCFE-UHFFFAOYNA-N
2-methoxy-2-phenylacetic acid
107202
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 230-256-

SDS
Documents

Documents

RUO – Research Use Only