missing translation for 'onlineSavingsMsg'
Learn More

Furosemide Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013884
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013884 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013884 Supplier Merck Emd Millipore Supplier No. PHR1816100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Furosemide Related Compound B is an impurity of the loop diuretic drug, furosemide. Furosemide is generally used to treat fluid retention in people with heart failure, liver cirrhosis or kidney disease.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 3086-91-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H7 ClN2O4 S
MDL Number MFCD03423689
Melting Point 267°C
Synonym 4-Chloro-5-sulfamoylanthranilic acid; 4-CHLORO-5- SULFAMOYLANTHRANILIC ACID
Type Certified Reference Material
Quantity 100 mg
Formula Weight 250.66
Grade Certified Reference Material