missing translation for 'onlineSavingsMsg'
Learn More

Furosemide Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014275
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014275 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014275 Supplier Merck Emd Millipore Supplier No. PHR1817100MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 4818-59-1
Linear Formula C12H11 ClN2O5 S
MDL Number MFCD18379506
Melting Point 197°C
Synonym 2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid; 2-CHLORO-4-N-FURFURYLAMINO -5-SULFAMOYLBENZOIC ACID
Type Certified Reference Material
Quantity 100 mg
Formula Weight 330.74
Grade Certified Reference Material