missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-boronic acid pinacol ester, 96%

Catalog No. p-7047258
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1836603 1 g
AAL1836606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1836603 Supplier Thermo Scientific Chemicals Supplier No. L1836603

missing translation for 'validMainframeMsg'

CAS: 374790-93-9 | C10H15BO3 | 194.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374790-93-9
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.04
MDL Number MFCD02094008
InChI Key GWEWQKZABZXLJH-UHFFFAOYSA-N
Synonym 2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol ester, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, furan-2-boronic acid pinacol cyclic ester, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, zlchem 693, pubchem18440
PubChem CID 11356013
IUPAC Name 2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CO1

Specifications

Refractive Index 1.472
Quantity 1 g
Beilstein 9045028
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material Furan-2-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only