missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fura-2, AM, FluoroPure™ grade - Special Packaging

Catalog No. F14185
Encompass
Change view
Click to view available options
Quantity:
20 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F14185 20 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F14185 Supplier Molecular Probes™ Supplier No. F14185
Only null left

Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade Fura-2 AM is greater than or equal to 98% pure by HPLC.

Because it is manufactured at our ISO 13485-certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade Fura-2 AM is greater than or equal to 98% pure by HPLC.

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Fluorescence Probe
For Use With (Equipment) Fluorescence Microscope
Product Type Fura-2
Dye Type Fluorescent Dye-Based
Product Line FluoroPure
Quantity 20 x 50 μg
Shipping Condition Room Temperature
I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

For Research Use Only. Not for use in diagnostic procedures.