missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Fura-2 AM Dye
Description
- Upon binding Ca2+, the excitation spectrum of Fura-2 shifts to shorter wavelengths between 300 and 400nm, while the peak emission remains steady around 510nm
- The Kd of Fura-2 is highly dependent on pH, temperature, ionic strength and viscosity of the cytosol, thus great care should be taken when non-standard conditions are used
- Molecular weight: 1001.86g/mol
- Peak excitation: variable depending on the concentration of free Ca2+, between 300 and 400nm
- Peak emission: 510nm
- Reconstitute in high-quality, freshly opened DMSO. Once reconstituted, protect from light and store at −20°C. Avoid freeze-thawing
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Product Line | eBioscience |
| Quantity | 1 mg |
| Emission Wavelength | 300 to 400/510 |
For Research Use Only. Not for use in diagnostic procedures.