missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fura-2, AM, cell permeant

Catalog No. F1201
Encompass
Change view
Click to view available options
Quantity:
20 x 50 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1201 1 mg
F1221 20 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1201 Supplier Molecular Probes™ Supplier No. F1201

Fura-2, AM, cell permeant

Fura-2, AM is a high affinity, intracellular calcium indicator that is ratiometric and UV light—excitable. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in a DMSO solution. Cell-impermeant pentapotassium and pentasodium salt forms of the dye are available.

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability, Proliferation and Cellular Imaging
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.