missing translation for 'onlineSavingsMsg'
Learn More

Fumonisin B1 reference material, MilliporeSigma™ Supelco™

Numéro de catalogue. 111012653
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111012653 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111012653 Fournisseur Merck Emd Millipore Code fournisseur 329365MG
Il en reste null

Fumonisins are a family of mycotoxins that are characterized by their structurally related homologs. They are generally produced by fungi of Fusarium species.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Spécifications

CAS 116355-83-0
Physical Form Neat
Linear Formula C34H59 NO15
MDL Number MFCD27977580
Synonym Fumonisin B1 from Fusarium moniliforme; Macrofusine
Type Analytical Standard
Quantity 5 mg
Formula Weight 721.83
Grade Analytical Standard