missing translation for 'onlineSavingsMsg'
Learn More

Fumaric acid, 99%

CAS: 110-17-8 | C4H4O4 | 116.07 g/mol

Supplier:  Thermo Scientific Chemicals A109760E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA109760E

Only null left

Description

Description

Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.

Notes
Incompatible with oxidizing agents, amines and strong bases.
Specifications

Chemical Identifiers

110-17-8
116.07
VZCYOOQTPOCHFL-OWOJBTEDSA-N
444972
(E)-but-2-enedioic acid
C4H4O4
MFCD00002700
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
CHEBI:18012
OC(=O)\C=C\C(O)=O

Specifications

Fumaric acid
∼299°C (sublimation)
2.1
Odorless
2500 g
605763
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(E)-but-2-enedioic acid
444972
116.07
110-17-8
1.625
273°C (523°F)
C4H4O4
MFCD00002700
14,4287
Soluble in water,ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform,olive oil,carbon tetrachloride,xylene,molten camphor,liquid ammonia and benzene.
OC(=O)\C=C\C(O)=O
116.07
CHEBI:18012
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319-H500

EINECSNumber : 203-743-0

RTECSNumber : LS9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions