missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaramide 96.0+%, TCI America™
$257.24 - $385.86
Chemical Identifiers
| CAS | 627-64-5 |
|---|---|
| Molecular Formula | C4H6N2O2 |
| Molecular Weight (g/mol) | 114.104 |
| MDL Number | MFCD00025527 |
| InChI Key | BSSNZUFKXJJCBG-OWOJBTEDSA-N |
| Synonym | fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide |
| PubChem CID | 446408 |
| IUPAC Name | (E)-but-2-enediamide |
| SMILES | C(=CC(=O)N)C(=O)N |
Chemical Identifiers
| 627-64-5 | |
| 114.104 | |
| BSSNZUFKXJJCBG-OWOJBTEDSA-N | |
| 446408 | |
| C(=CC(=O)N)C(=O)N |
| C4H6N2O2 | |
| MFCD00025527 | |
| fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide | |
| (E)-but-2-enediamide |
Specifications
| 627-64-5 | |
| C4H6N2O2 | |
| 10 g | |
| BSSNZUFKXJJCBG-OWOJBTEDSA-N | |
| (E)-but-2-enediamide | |
| 446408 | |
| ≥96.0% (N) | |
| Fumaramide |
| White | |
| MFCD00025527 | |
| fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide | |
| C(=CC(=O)N)C(=O)N | |
| 114.104 | |
| 114.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : No