accessibility menu, dialog, popup

UserName

CAS RN 627-64-5

NH2ONH2O(E)

CAS RN 627-64-5

IUPAC Name: (2E)-but-2-enediamide
Synonyms: 2-butenediamide, (E)-
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: BSSNZUFKXJJCBG-OWOJBTEDSA-N
SMILES: NC(=O)\C=C\C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results