missing translation for 'onlineSavingsMsg'
Learn More

Fumaraldehyde Mono(dimethyl Acetal) (stabilized with HQ) 95.0+%, TCI America™

Catalog No. F03505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03505G 5 g
F03501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03505G Supplier TCI America Supplier No. F03505G
Only null left

Chemical Identifiers

CAS 4093-49-6
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00191439
InChI Key OJFHAFJGQZSFKT-ONEGZZNKSA-N
Synonym 4,4-Dimethoxy-2-butenal, 4,4-Dimethoxycrotonaldehyde
PubChem CID 10725377
IUPAC Name (E)-4,4-dimethoxybut-2-enal
SMILES COC(C=CC=O)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Fumaraldehyde Mono(dimethyl Acetal) (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator