accessibility menu, dialog, popup

UserName

CH3OOOH3C(E/Z)

CAS RN 4093-49-6

IUPAC Name: 4,4-dimethoxybut-2-enal
Synonyms: 4,4-dimethoxybut-2-enal
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: OJFHAFJGQZSFKT-UHFFFAOYSA-N
SMILES: COC(OC)C=CC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results