missing translation for 'onlineSavingsMsg'
Learn More

FuGENE™ 4K Transfection Reagent

Catalog No. PRE5911
Encompass
Change view
Click to view available options
:
Cell Type: Mammalian
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRE5911 Cell Type: Mammalian
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE5911 Supplier FuGENE™ Supplier No. E5911
Only null left

Achieve high-efficiency transfection in challenging and routine cell lines with the most effective broad-spectrum DNA transfection reagent. It is gentle on cell viability, making it ideal for protein and virus production.

  • Effective transfection reagent for a variety of cell lines.
  • High transfection efficiency in both challenging and routine cell lines.
  • Minimizes impact on cell viability.
  • Ideal for protein and virus production.
  • Gentle on cells, ensuring higher viability.
  • Consistent performance across different experiments.
  • Suitable for various applications, including gene expression studies.
  • Easy to use with a simple protocol.
  • Produces reliable and reproducible results.
  • Cost-effective solution for transfection needs.

Specifications

Cell Type Mammalian
Format Bottle
Transfection Technique Chemical Transfection
Product Type Reagent