missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucoxanthin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 3351-86-8 |
| Physical Form | Neat |
| Linear Formula | C42H58O6 |
| MDL Number | MFCD01745140 |
| Synonym | all-trans-Fucoxanthin |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 658.91 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |