missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSH beta Monoclonal Antibody (C5)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566092
Description
FSH beta Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids.Chemical Identifiers
| FSH beta | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P01225, P01228, P18427 | |
| 2488, 25447, 396895 | |
| Recombinant Follicle Stimulating Hormone Beta (FSHb) (Ser21-Glu130). | |
| Antibody |
| C5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Rat, Pig | |
| FSHB | |
| follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24 | |
| FSHB | |
| Primary | |
| Affinity chromatography |
Specifications
| FSH beta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P01225, P01228, P18427 | |
| FSHB | |
| Recombinant Follicle Stimulating Hormone Beta (FSHb) (Ser21-Glu130). | |
| 200 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C5 | |
| Unconjugated | |
| FSHB | |
| follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2488, 25447, 396895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |