missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructose, Granular, USP, 98-102%, Spectrum™ Chemical
FR106, 57-48-7, C6H12O6
Supplier: Spectrum Chemical Mfg Cor FR10612KG
Description
Spectrum™ Chemical Fructose, Granular, USP is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used as a sweetener in pharmaceutical formulations. Spectrum Chemical manufactured USP FRUCTOSE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 57-48-7 | |
| 0.5% | |
| C6H12O6 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
| 98 to 102% |
| 100% | |
| Poly Pail | |
| 12 kg | |
| OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO | |
| 180.16 | |
| USP |
Chemical Identifiers
| 57-48-7 | |
| 180.16 | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
| C6H12O6 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO |