missing translation for 'onlineSavingsMsg'
Learn More

Fructose, Granular, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7157478
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602862 2.5 kg Poly Bottle
18602861 6 x 500 g Poly Bottle
18602863 4 x 2.5 kg Poly Bottle
18602864 12 kg Poly Pail
18602865 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602862 Supplier Spectrum Chemical Mfg Cor Supplier No. FR1062.5KG
Only null left

FR106, 57-48-7, C6H12O6

Spectrum™ Chemical Fructose, Granular, USP is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used as a sweetener in pharmaceutical formulations. Spectrum Chemical manufactured USP FRUCTOSE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98 to 102%
Grade USP