missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructose, 2000ppm, Ricca Chemical
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Min % | 98.0, 0.2 |
| CAS Max % | 100.0, 0.2 |
| Chemical Name or Material | Fructose Standard, 2000ppm |