missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRS2 Recombinant Rabbit Monoclonal Antibody (23GB5785)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552916
Description
FRS2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1. [UniProt].Specifications
| FRS2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FRS2 | |
| 4732458E18; C330018A15Rik; FGFR signalling adaptor; FGFR substrate 2; FGFR-signaling adaptor SNT; fibroblast growth factor receptor substrate 2; FRS1A; FRS2; Frs2a; FRS2alpha; FRS2-alpha; SNT; SNT1; SNT-1; SNT2; suc1-associated neurotrophic factor target 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10818 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5785 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WU20 | |
| FRS2 | |
| A synthesized peptide derived from human FRS2 (100-250AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |