missing translation for 'onlineSavingsMsg'
Learn More

Ohaus Frontier™ 5513 Micro Centrifuge with 24 x 1.5/2.0 L Rotor

Catalog No. 01675095
Change view
Click to view available options
Capacity:
24 x 1.5/2.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
01675095 24 x 1.5/2.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01675095 Supplier Ohaus Supplier No. 30370692

Frontier™ Micro centrifuges deliver high-speed operation for a range of essential lab applications, such as DNA/RNA preparation, ultrafiltration, and more. Centrifuge features an intuitive interface, with all key parameters immediately accessible for observation and adjustment.

  • Frontier 5513 offers standard 24 x 1.5/2.0 mL microtube rotors with a biosealed option
  • Centrifuge can also support spin columns and hematocrit capillary tubes when used with additional rotor options
  • Features convenient control on a protective splash-proof interface, which allows for responsive operation - even when wearing gloves.
  • Frontier 5513 centrifuges are compact enough to save valuable space on the bench, yet large enough to accommodate the most common size 24 x 1.5/2.0 mL rotors
  • One-touch toggling between RPM/RCF, Acc/Dec
  • Crisp display on a double-line LCD display with backlight
  • Maintenance-free induction motor produces maximum G-force of 17,317 x g
  • Speeds up to 13,500 rpm in less than 15 seconds with 24 x 1.5/2.0 mL rotor
  • 10 acceleration/deceleration settings
  • 99 memory sets to quickly store and recall commonly used parameters
  • Rotor imbalance sensing systems stops the unit when it detects an imbalanced load

Specifications

Capacity 24 x 1.5/2.0 mL
Max. Speed 13,500 rpm
Max. RCF 17,317 x g
Refrigerated No
Voltage 120 V
Product Type Microcentrifuge