Learn More
Thermo Scientific™ Storage Boxes
Store a variety of vials and tubes with these fiberboard storage boxes, for use in ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 820003
 View more versions of this product
Catalog No. 15465462
Only null left
Description
Description
Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.
  • Sturdy, water-repellant boxes with covers for sample storage
  • Available in 2 and 3 in. high (5.1 and 7.6cm) with 5.25 x 5.25 in. (13.3 x 13.3cm) square bases
  • Box dividers included (excluding 5956)
Specifications
Chemical Identifiers
White
Square
Cryo Storage Box
Thermo Scientific Ultra-Low Temperature Freezers
Specifications
White
Square
Thermo Scientific Ultra-Low Temperature Freezers
Dividers
5 in.
5 in.
3 in.
0.08 kg
Fiberboard
Cryo Storage Box
81 Cryo Tubes
5 x 5 x 3 in.
12.7 cm
12.7 cm
8 cm
Product Suggestions
Product Suggestions