missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hughes Freeze Protected, Floor Mount Drench Shower

Description
Features:
- Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength.
- Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions.
- Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur.
- Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
- Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
- Shower features a removable, self-draining overhead nozzle designed for high flow rates that meet or exceed ANSI standards.
- Made of durable nylon material, the nozzle offers optimal spray distribution for user protection.
- Includes an inspection record tag for recording service and test history.
- Electrical supply: 120V Single Phase, 60/50Hz
- For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
- Suitable for both Class 1 Division 2 hazardous areas, as well as non-hazardous areas
- Includes a universal safety shower sign for enhanced visibility
- Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Compliance
ANSI Z358.1-2014 and EN15154 Compliant
Warranty
1 Year
Specifications
Specifications
| Activation | Pull Handle |
| Certifications/Compliance | ANSI Z358.1-2014 and EN15154 Compliant |
| Color | White with Green Stripes |
| Depth (English) | 16 in. |
| Depth (Metric) | 406 mm |
| Electrical Requirements | 120 V |
| For Use With (Application) | Emergency Shower and Eye Wash with Water |
| Height (English) | 48 in. |
| Height (Metric) | 1219 mm |
| Includes | 1 Unit |
| Show More |