missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Free Standing Eyewash and Shower Station

Catalog No. 06000439
Change view
Click to view available options
For Use With (Application):
ADA Safety Shower
Safety Shower
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
06000439 Safety Shower
06000440 ADA Safety Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06000439

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Free Standing Eyewash and Shower Station includes a drench shower and eye wash fixture.

  • Drench shower is made out of ABS plastic
  • Ball valve operated by rigid pull-rod and easy grip handle
  • Eye wash fixture with stainless steel bowl, four gently spray heads and a Push to Operate ball valve
  • Eye wash and shower operate independently
  • Overall height 80 in.

Specifications

Color Orange
Flow Rate 20 gpm
For Use With (Application) Safety Shower
Includes Free-standing Drench Shower and Eyewash
Material ABS Plastic
Mounting Floor
Sterility Non-sterile