Learn More
National Safety Apparel™ FR Chemical Resistant Lab Coat, Navy Blue
Chemical resistant FR lab coat features proprietary CR/FR fabric to provide multi-hazard protection without weighing down the wearer.
$513.02 - $513.02
Specifications
Product Type Flame-resistant Lab Coat
Gender Unisex
Material Aramid Fiber
Color Navy
Closure Type Snaps
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17000308
View Documents
National Safety Apparel™
LABCRNPNBMD
Medium
Each for $513.02
Only null left
Description
Description
  • FR Nomex™ knit wrists keep sleeves in place
  • Durable, high-temp plastic snaps for easy donning and doffing
  • Bottom side vents for increased mobility
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Chest pocket with covered flap and pen/pencil slot
  • Lower pockets with covered flaps to secure contents
  • Highly breathable compared to traditional chemical protective options
  • Easy care and laundering
  • 42 in. length
Specifications
Specifications
Flame-resistant Lab Coat
Aramid Fiber
Snaps
Reusable
Fluids, Flame
3
Unisex
Navy
Knit
Non-sterile
Standard Length
NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'