missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ FR Chemical Resistant Lab Coat, Navy Blue

Catalog No. 17000308
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17000308 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000308 Supplier National Safety Apparel™ Supplier No. LABCRNPNBMD
Only null left

Chemical resistant FR lab coat features proprietary CR/FR fabric to provide multi-hazard protection without weighing down the wearer.

  • FR Nomex™ knit wrists keep sleeves in place
  • Durable, high-temp plastic snaps for easy donning and doffing
  • Bottom side vents for increased mobility
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Chest pocket with covered flap and pen/pencil slot
  • Lower pockets with covered flaps to secure contents
  • Highly breathable compared to traditional chemical protective options
  • Easy care and laundering
  • 42 in. length

Specifications

Product Type Flame-resistant Lab Coat
Size Medium
Gender Unisex
Material Aramid Fiber
Color Navy
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Flame
Style Standard Length
No. of Pockets 3
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
For Use With (Application) Manufacturing, Food Processing
Show More Show Less