missing translation for 'onlineSavingsMsg'
Learn More

Foxp3 Rat, PE, Clone: PCH101, Staining Set, eBioscience™

Catalog No. 5016976
Change view
Click to view available options
Quantity:
Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5016976 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5016976 Supplier Thermo Fisher Scientific Supplier No. 72577640

Includes

  • Fixation/Permeabilization Concentrate (4X) (cat. 00-5123): 30mL, store at 2-8°C. Avoid agitation.
  • Fixation/Permeabilization Diluent (cat. 00-5223): 100mL, store at 2-8°C.
  • Permeabilization Buffer (10X) (cat. 00-8333): 100mL, store at 2-8°C. Note: The 10X Permeabilization Buffer has a natural tendency to precipitate, however, its function is not affected by this. To clarify, the solution can be filtered after dilution to 1x working solution.
  • Normal Rat Serum (cat. 24-5555): 100 μL, store at 2-8°C.
  • Anti-Human Foxp3 PE (clone PCH101, cat. 12-4776): 25 tests, store at 2-8°C. Light-sensitive material.

Antibody Staining Kit

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3+ cells. The PCH101 monoclonal antibody reacts with the amino terminus of human Foxp3, also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The PCH101 antibody crossreacts with rhesus, chimpanzee, and cynomolgus Foxp3. PCH101 recognizes a different epitope of Foxp3 than clones 236A/E7 and 150D/E4.

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone PCH101
Conjugate PE
Gene Alias Forkhead Box P3, Scurfin, JM2
Host Species Rat
Quantity Kit
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chimpanzee, Cynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
Content And Storage Store at 2-8°C. Do not freeze. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.