missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anti-Human Foxp3 Staining Set PE, eBioscience™

Catalog No. 5016974
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5016974 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5016974 Supplier Invitrogen™ Supplier No. 72577440

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Fixation/Permeabilization Concentrate (4X) (cat. 00-5123): 30mL, store at 2-8°C. Avoid agitation.
  • Fixation/Permeabilization Diluent (cat. 00-5223): 100mL, store at 2-8°C.
  • Permeabilization Buffer (10X) (cat. 00-8333): 100mL, store at 2-8°C. Note: The 10X Permeabilization Buffer has a natural tendency to precipitate, however, its function is not affected by this. To clarify, the solution can be filtered after dilution to 1x working solution.
  • Normal Mouse Serum (cat. 24-5544): 100 μL, store at 2-8°C.
  • Anti-Human Foxp3 PE (clone 236A/E7, cat. 12-4777): 25 tests, store at 2-8°C. Light-sensitive material.

Antibody Staining Kit

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3+ cells. The 236A/E7 monoclonal antibody reacts with human Foxp3, also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The 236A/E7 antibody has been shown to crossreact with rhesus macaque, sooty mangabey, and cynomolgus Foxp3. 236A/E7 recognizes a different epitope of Foxp3 than PCH101 and 150D/E4.

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone 236A/E7
Conjugate PE
Gene Alias Forkhead Box P3, Scurfin, JM2
Host Species Mouse
Quantity 1 kit
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
Content And Storage Store at 2-8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
Isotype IgG1
Product Line eBioscience
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.