missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXP1 Recombinant Human Monoclonal Antibody (RAB-S37)
GREENER_CHOICE

Catalog No. MA551872
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551872 Supplier Invitrogen™ Supplier No. MA551872
Only null left

Human Recombinant Monoclonal Antibody

This antibody recognizes FOXP1 (Forkhead box protein P1). It binds to a folded domain, amino acids 302-541. FOXP1 is a transcriptional repressor that plays an important role in the specification and differentiation of lung epithelium.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen FOXP1
Applications ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RAB-S37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene FOXP1
Gene Accession No. Q9H334
Gene Alias 12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; Fork head related protein like B; fork head-related protein like B; forkhead box P1; forkhead box protein P1; Forkhead box protein P1 (FOXP1); Forkhead-related transcription factor 1; Foxp1; Glutamine rich factor 1; glutamine-rich factor 1; hFKH1B; HSPC215; mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1
Gene Symbols FOXP1
Host Species Human
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP1 protein under non-denaturing conditions.
Purification Method Metal-chelate chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype F(ab) κ
Show More Show Less