missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD4, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002298B01P
Description
Sequence: MNLPRAERLRSTPQRSLRDSDGEDGKIDVLGEEEDEDEEEAASQQFLEQSLQPGLQVARWGGVALPREHIEGGGGPSDPSEFGTEFRAPPRSAAASEDARQPAKPPSSYIALITMAILQSPHKRLTLSGICAFISDRFPYYRRKFPAWQNSIRHNLSLNDCFVKIPREPGRPGKGNYWSLDPASQDMFDNGSFLRRRKRFQRHQPTPGAHLPHPFPLPAAHAALHNPRPGPLLGAPAPPQPVPGAYPNTGPGRRPYALLHPHPPRYLLLSAPAYAGAPKKAEGADLATPAPFPCCSPHLVLSLGRRARVWRRHREADASLSALRVSCKGSGERVQGLRRVCPRPRGATAPCSSDRQACRTILQQQQRHQEEDCANGCAPTKGAVLGGHLSAASALLRYQAVAEGSGLTSLAAPLGGEGTSPVFLVSPTPSSLAESAGPSSpecifications
| FOXD4 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_207305.2 | |
| FOXD4 | |
| FOXD4 (NP_997188.2, 1 a.a. ∼ 439 a.a) full-length human protein. | |
| RUO | |
| 2298 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| forkhead box D4 | |
| FKHL9/FOXD4A/FREAC5/MGC105106 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |