missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin Disodium Salt 98.0+%, TCI America™

Catalog No. F088925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F088925G 25 g
F08895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F088925G Supplier TCI America Supplier No. F088925G

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3H5Na2O4P
Molecular Weight (g/mol) 182.02
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
Synonym Phosphomycin Disodium Salt, Disodium (1R,2S)-1,2-Epoxypropylphosphonate, (1R,2S)-1,2-Epoxypropylphosphonic Acid Disodium Salt
PubChem CID 16397688
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

Specifications

Type Fosfomycin Disodium Salt
Physical Form Crystalline Powder
Color White
Quantity 25 g
Formula Weight 182.02
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Fosfomycin Disodium Salt
RTECSNumber SZ7902000
TSCA No
Recommended Storage Refrigerator