missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin disodium, 90%

Catalog No. AAJ6660206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6660206 5 g
AAJ6660214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6660206 Supplier Thermo Scientific Chemicals Supplier No. J6660206*

Prevents peptidoglycan synthesis by inhibiting MurA | CAS: 26016-99-9 | C3H5Na2O4P | 182.02 g/mol

Prevents peptidoglycan synthesis by inhibiting MurA, an enzyme responsible for synthesizing N-acetylmuramic acid or NAM, a major component of peptidoglycan

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3H5Na2O4P
Molecular Weight (g/mol) 182.02
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
PubChem CID 16397688
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

Specifications

Type Fosfomycin Disodium
Physical Form Powder
Melting Point >300°C
Color White
Quantity 5 g
Beilstein 4604425
Synonym Phosphomycin disodium salt, (-)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, disodium salt
Solubility Information Freely soluble in water or aqueous solution
Formula Weight 182.02
Percent Purity 90%
Chemical Name or Material Fosfomycin disodium
Show More Show Less
EINECSNumber 247-409-2
RTECSNumber SZ7902000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only