missing translation for 'onlineSavingsMsg'
Learn More

Foscarnet Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013972
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013972 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013972 Supplier Merck Emd Millipore Supplier No. PHR14361G
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 34156-56-4 (anhydrous)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula CNa3O5 P · 6H2 O
Synonym Foscarnet sodium hexahydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 300.04
Grade Certified Reference Material