missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forskolin 95.0+%, TCI America™
$273.39 - $922.72
Chemical Identifiers
| CAS | 66575-29-9 |
|---|---|
| Molecular Formula | C22H34O7 |
| Molecular Weight (g/mol) | 410.507 |
| MDL Number | MFCD00082317 |
| InChI Key | OHCQJHSOBUTRHG-KGGHGJDLSA-N |
| Synonym | forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish |
| PubChem CID | 47936 |
| ChEBI | CHEBI:42471 |
| IUPAC Name | [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
| SMILES | CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
Chemical Identifiers
| 66575-29-9 | |
| 410.507 | |
| OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
| 47936 | |
| [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
| C22H34O7 | |
| MFCD00082317 | |
| forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
| CHEBI:42471 | |
| CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
Specifications
| 66575-29-9 | |
| White | |
| MFCD00082317 | |
| OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
| [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate | |
| 47936 | |
| 410.51 | |
| Crystalline Powder |
| 225°C | |
| C22H34O7 | |
| forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
| CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O | |
| 410.507 | |
| CHEBI:42471 | |
| ≥95.0% (HPLC) | |
| Forskolin |
Safety and Handling
RTECSNumber : QL6150000
TSCA : No