missing translation for 'onlineSavingsMsg'
Learn More

Forskolin 95.0+%, TCI America™

Catalog No. F085550MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F085550MG 50 mg
F085510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F085550MG Supplier TCI America Supplier No. F085550MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
MDL Number MFCD00082317
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Melting Point 225°C
Color White
Quantity 50 mg
Formula Weight 410.51
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Forskolin
RTECSNumber QL6150000
TSCA No