missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Formononetin | |
| 258°C | |
| MFCD00016948 | |
| formononetin, biochanin b, formononetol, 7-hydroxy-4'-methoxyisoflavone, 7-hydroxy-3-4-methoxyphenyl-4h-chromen-4-one, 4'-o-methyldaidzein, 7-hydroxy-3-4-methoxyphenyl chromen-4-one, unii-295dqc67bj, 7-hydroxy-3-4-methoxyphenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-methoxyphenyl | |
| COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O | |
| 268.27 | |
| CHEBI:18088 | |
| ≥98.0% (HPLC,T) |
| 485-72-3 | |
| C16H12O4 | |
| 1 g | |
| HKQYGTCOTHHOMP-UHFFFAOYSA-N | |
| 7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one | |
| 5280378 | |
| 268.27 | |
| Crystalline Powder |
Chemical Identifiers
| 485-72-3 | |
| 268.27 | |
| HKQYGTCOTHHOMP-UHFFFAOYSA-N | |
| 5280378 | |
| 7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one |
| C16H12O4 | |
| MFCD00016948 | |
| formononetin, biochanin b, formononetol, 7-hydroxy-4'-methoxyisoflavone, 7-hydroxy-3-4-methoxyphenyl-4h-chromen-4-one, 4'-o-methyldaidzein, 7-hydroxy-3-4-methoxyphenyl chromen-4-one, unii-295dqc67bj, 7-hydroxy-3-4-methoxyphenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-methoxyphenyl | |
| CHEBI:18088 | |
| COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O |
Safety and Handling
RTECSNumber : DJ3100114
TSCA : No