missing translation for 'onlineSavingsMsg'
Learn More

Formononetin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014287
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014287 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014287 Supplier Merck Emd Millipore Supplier No. 9433450MG

missing translation for 'validMainframeMsg'

Formononetin belongs to the isoflavones class of polyphenols.

Featured Industries: Food and Beverages

Specifications

CAS 485-72-3
Linear Formula C16H12O4
MDL Number MFCD00016948
Synonym 7-Hydroxy-3-(4-methoxyphenyl)-4 H-1-benzopyran-4-one; 7-Hydroxy-3-(4-methoxyphenyl)chromone; 7-Hydroxy-4′-methoxyisoflavone; Biochanin B; Daidzein 4′-methyl ether; Formonetin; Formononetol
Type Analytical Standard
Quantity 50 mg
Formula Weight 268.26
Percent Purity ≥98% (HPLC)
Grade Analytical Standard