Learn More
StatLab™ Formical-4 Decalcifier
Decalcifies large specimens, allowing better penetration of formaldehyde.
Supplier:  StatLab™ 12141
 View more versions of this product
Catalog No. STL12141
Only null left
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Description
Description
  • Fresh tissue can be placed directly into Formical-4 to fix and decalcify simultaneously
  • Formulated for use on large dense samples that normally take several days to decalcify
  • Works well for IHC and ISH
Specifications
Specifications
4 x 1 gal.
Decalcifier
Product Suggestions
Product Suggestions