Learn More
StatLab™ Formical-4 Decalcifier
Decalcifies large specimens, allowing better penetration of formaldehyde.
$199.79 - $444.54
Specifications
Solution Type Decalcifier
View More Specs
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
STL12141
View Documents Promotion Details
StatLab™
12141
4 x 1 gal.
Each for $199.79
Case of 4 Each for $799.16
Only null left
STL12145
View Documents Promotion Details
StatLab™
12145
5 gal. Cube
Each for $444.54
Only null left
Description
Description
  • Fresh tissue can be placed directly into Formical-4 to fix and decalcify simultaneously
  • Formulated for use on large dense samples that normally take several days to decalcify
  • Works well for IHC and ISH
Specifications
Specifications
Decalcifier