Learn More
StatLab™ Formical-4 Decalcifier
Description
- Fresh tissue can be placed directly into Formical-4 to fix and decalcify simultaneously
- Formulated for use on large dense samples that normally take several days to decalcify
- Works well for IHC and ISH
Spécifications
Spécifications
| Quantity | 5 gal. Cube |
| Solution Type | Decalcifier |