Learn More
StatLab™ Formical-2000 Decalcifier
A fast yet gentle bone decalcifier.
Supplier:  StatLab™ 13141
 Afficher plus de versions de ce produit
Numéro de catalogue. STL13141
Il en reste null
Découvrez plus d'offres spéciales
Avertissement:
Harmful if swallowed, in contact with skin, or inhaled.
Description
Description
  • Formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA
  • Works for all routine histological specimens, as well as IHC
  • Will safely decalcify bone without destroying antigen sites
Spécifications
Spécifications
1 Gallon
Decalcifier
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS